Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CCDC57
Entrez Gene ID
284001
Gene Name
coiled-coil domain containing 57
Gene Aliases
-
Chromosome Location
Chr.17: 82101458 - 82212892 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001316321.1 NP_001303250.1 3 - 4 780 119
NM_198082.2 NP_932348.2 2 - 3 446 119
XM_011523545.2 XP_011521847.2 3 - 4 683 119
XM_011523546.2 XP_011521848.1 2 - 3 1015 119
XM_011523548.2 XP_011521850.1 3 - 4 711 119
XM_011523549.2 XP_011521851.1 2 - 3 525 119
XM_011523551.2 XP_011521853.1 3 - 4 724 119
XM_011523556.2 XP_011521858.2 3 - 4 677 119
XM_017024462.1 XP_016879951.1 3 - 4 685 119
XM_017024463.1 XP_016879952.1 3 - 4 674 119
XM_017024464.1 XP_016879953.1 3 - 4 676 119
XM_017024465.1 XP_016879954.1 3 - 4 675 119
XM_017024466.1 XP_016879955.1 3 - 4 664 119
XM_017024467.1 XP_016879956.1 3 - 4 840 119
XM_017024468.1 XP_016879957.1 3 - 4 671 119
XM_017024472.1 XP_016879961.1 3 - 4 724 119
XM_017024473.1 XP_016879962.1 3 - 4 670 119
XM_017024474.1 XP_016879963.1 3 - 4 674 119
XM_017024476.1 XP_016879965.1 2 - 3 458 119
XM_017024479.1 XP_016879968.1 3 - 4 710 119
XM_017024480.1 XP_016879969.1 3 - 4 683 119
XM_017024481.1 XP_016879970.1 3 - 4 683 119
XM_017024482.1 XP_016879971.1 3 - 4 687 119
GenBank mRNA BC040264.1 - 3 - 4 764 119
BC110997.1 - 2 - 3 446 119

View Full Product Details