The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001323623.1 | NP_001310552.1 | 1 - 2 | 224 | 78 |
NM_001323624.1 | NP_001310553.1 | 1 - 2 | 224 | 78 | |
NM_013285.2 | NP_037417.1 | 1 - 2 | 224 | 78 | |
GenBank mRNA | AK298358.1 | - | 1 - 2 | 178 | 78 |
AK307352.1 | - | 1 - 2 | 178 | 78 | |
AK312915.1 | - | 1 - 2 | 178 | 78 | |
BC000107.2 | - | 1 - 2 | 160 | 78 | |
BC009250.2 | - | 1 - 2 | 160 | 78 | |
DB452094.1 | - | 1 - 2 | 224 | 78 | |
L05425.1 | - | 1 - 2 | 140 | 78 |