Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CTSA
Entrez Gene ID
5476
Gene Name
cathepsin A
Gene Aliases
GLB2, GSL, NGBE, PPCA, PPGB
Chromosome Location
Chr.20: 45890144 - 45898820 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000308.3 NP_000299.2 8 - 9 2007 83
NM_001127695.2 NP_001121167.1 8 - 9 2053 83
NM_001167594.2 NP_001161066.1 7 - 8 1956 83
NR_133656.1 8 - 9 2014 83
GenBank mRNA AB209705.2 - 8 - 9 823 83
AK097786.1 - 7 - 8 720 83
AK172808.1 - 8 - 9 2011 83
AK304536.1 - 7 - 8 784 83
AK307701.1 - 8 - 9 800 83
AK312898.1 - 8 - 9 816 83
BC000597.2 - 8 - 9 809 83
BC093009.1 - 8 - 9 816 83
M22960.1 - 7 - 8 777 83

View Full Product Details