The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001681.3 | NP_001672.1 | 12 - 13 | 2112 | 69 |
NM_170665.3 | NP_733765.1 | 12 - 13 | 2112 | 69 | |
XM_005253888.2 | XP_005253945.1 | 12 - 13 | 2107 | 69 | |
XM_011538402.2 | XP_011536704.1 | 12 - 13 | 2107 | 69 | |
GenBank mRNA | AK293877.1 | - | 8 - 9 | 1220 | 69 |
BC035588.1 | - | 12 - 13 | 1659 | 69 | |
KU177929.1 | - | 12 - 13 | 1549 | 69 | |
M23114.1 | - | 12 - 13 | 1712 | 69 | |
M23115.1 | - | 12 - 13 | 1552 | 69 |