The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001319066.1 | NP_001305995.1 | 5 - 6 | 727 | 154 |
NM_001319067.1 | NP_001305996.1 | 5 - 6 | 629 | 154 | |
NM_001319068.1 | NP_001305997.1 | 5 - 6 | 600 | 154 | |
NM_001319069.1 | NP_001305998.1 | 3 - 4 | 429 | 154 | |
NM_004763.4 | NP_004754.1 | 5 - 6 | 615 | 154 | |
NR_134951.1 | 5 - 6 | 604 | 154 | ||
NR_134956.1 | 4 - 5 | 536 | 154 | ||
NR_134957.1 | 4 - 5 | 508 | 154 | ||
XM_005246183.4 | XP_005246240.1 | 6 - 7 | 1301 | 154 | |
XM_005246184.4 | XP_005246241.1 | 6 - 7 | 740 | 154 | |
XM_005246185.4 | XP_005246242.1 | 6 - 7 | 613 | 154 | |
XM_006711903.3 | XP_006711966.1 | 5 - 6 | 845 | 154 | |
XM_011510416.2 | XP_011508718.1 | 6 - 7 | 935 | 154 | |
XM_017005267.1 | XP_016860756.1 | 6 - 7 | 2430 | 154 | |
XM_017005268.1 | XP_016860757.1 | 6 - 7 | 605 | 154 | |
GenBank mRNA | AF012023.1 | - | 5 - 6 | 551 | 154 |
AK289817.1 | - | 5 - 6 | 528 | 154 | |
AK299017.1 | - | 3 - 4 | 322 | 154 | |
BC012264.1 | - | 5 - 6 | 507 | 154 | |
BM823938.1 | - | 4 - 5 | 335 | 154 | |
BU520903.1 | - | 5 - 6 | 574 | 154 | |
BX647099.1 | - | 4 - 5 | 468 | 154 | |
CR457006.1 | - | 4 - 5 | 383 | 154 | |
CT002942.1 | - | 4 - 5 | 416 | 154 | |
DB167886.1 | - | 5 - 6 | 510 | 154 | |
DC321859.1 | - | 3 - 4 | 316 | 154 | |
DC361372.1 | - | 3 - 4 | 432 | 154 |