The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002831.5 | NP_002822.2 | 4 - 5 | 756 | 61 |
NM_080548.4 | NP_536858.1 | 4 - 5 | 669 | 61 | |
NM_080549.3 | NP_536859.1 | 4 - 5 | 756 | 61 | |
XM_006718994.1 | XP_006719057.1 | 4 - 5 | 813 | 61 | |
XM_011520988.1 | XP_011519290.1 | 4 - 5 | 670 | 61 | |
GenBank mRNA | AF178946.1 | - | 4 - 5 | 538 | 61 |
AK223629.1 | - | 4 - 5 | 651 | 61 | |
AK290421.1 | - | 4 - 5 | 639 | 61 | |
AK298470.1 | - | 3 - 4 | 922 | 61 | |
AK300167.1 | - | 3 - 4 | 513 | 61 | |
AK308230.1 | - | 4 - 5 | 704 | 61 | |
AK313379.1 | - | 4 - 5 | 669 | 61 | |
BC002523.2 | - | 4 - 5 | 655 | 61 | |
BC007667.2 | - | 4 - 5 | 655 | 61 | |
BG754792.1 | - | 4 - 5 | 689 | 61 | |
BT007390.1 | - | 3 - 4 | 520 | 61 | |
M74903.1 | - | 4 - 5 | 597 | 61 | |
M77273.1 | - | 4 - 5 | 658 | 61 |