The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006851.2 | NP_006842.2 | 1 - 2 | 323 | 66 |
XM_011537775.2 | XP_011536077.1 | 1 - 2 | 302 | 66 | |
GenBank mRNA | AF400440.1 | - | 1 - 2 | 203 | 66 |
AK304847.1 | - | 1 - 2 | 303 | 66 | |
AK313449.1 | - | 1 - 2 | 189 | 66 | |
BC012510.1 | - | 1 - 2 | 239 | 66 | |
CB123261.1 | - | 1 - 2 | 376 | 66 | |
EF667354.1 | - | 1 - 2 | 175 | 66 | |
U16307.3 | - | 1 - 2 | 259 | 66 | |
X91911.1 | - | 1 - 2 | 303 | 66 |