The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001271769.1 | NP_001258698.1 | 17 - 18 | 2160 | 120 |
NM_003664.4 | NP_003655.3 | 17 - 18 | 2148 | 120 | |
XM_005248618.3 | XP_005248675.1 | 17 - 18 | 2120 | 120 | |
XM_005248619.4 | XP_005248676.1 | 17 - 18 | 2120 | 120 | |
XM_017010001.1 | XP_016865490.1 | 17 - 18 | 1900 | 120 | |
GenBank mRNA | AK291201.1 | - | 17 - 18 | 2089 | 120 |
AY623424.1 | - | 17 - 18 | 2050 | 120 | |
BC038444.1 | - | 17 - 18 | 2109 | 120 | |
BX538041.1 | - | 17 - 18 | 2098 | 120 | |
KU178426.1 | - | 17 - 18 | 1972 | 120 | |
KU178427.1 | - | 17 - 18 | 1972 | 120 | |
U81504.1 | - | 17 - 18 | 2064 | 120 | |
U91931.1 | - | 17 - 18 | 2025 | 120 |