The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001271769.1 | NP_001258698.1 | 24 - 25 | 3083 | 124 |
NM_003664.4 | NP_003655.3 | 24 - 25 | 3071 | 124 | |
XM_005248618.3 | XP_005248675.1 | 24 - 25 | 3043 | 124 | |
XM_017010001.1 | XP_016865490.1 | 24 - 25 | 2823 | 124 | |
GenBank mRNA | AK225908.1 | - | 24 - 25 | 2947 | 124 |
AK291201.1 | - | 24 - 25 | 3012 | 124 | |
BC038444.1 | - | 24 - 25 | 3032 | 124 | |
BX538041.1 | - | 24 - 25 | 3021 | 124 | |
KU178426.1 | - | 24 - 25 | 2895 | 124 | |
KU178427.1 | - | 23 - 24 | 2788 | 124 | |
U81504.1 | - | 24 - 25 | 2987 | 124 | |
U91931.1 | - | 24 - 25 | 2945 | 124 |