The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001271769.1 | NP_001258698.1 | 4 - 5 | 564 | 106 |
NM_003664.4 | NP_003655.3 | 4 - 5 | 552 | 106 | |
XM_005248618.3 | XP_005248675.1 | 4 - 5 | 524 | 106 | |
XM_005248619.4 | XP_005248676.1 | 4 - 5 | 524 | 106 | |
XM_017010001.1 | XP_016865490.1 | 4 - 5 | 304 | 106 | |
GenBank mRNA | AK225908.1 | - | 4 - 5 | 428 | 106 |
AK291201.1 | - | 4 - 5 | 493 | 106 | |
AL708147.1 | - | 4 - 5 | 514 | 106 | |
AY623424.1 | - | 4 - 5 | 454 | 106 | |
BC038444.1 | - | 4 - 5 | 513 | 106 | |
BX538041.1 | - | 4 - 5 | 502 | 106 | |
KU178426.1 | - | 4 - 5 | 376 | 106 | |
KU178427.1 | - | 4 - 5 | 376 | 106 | |
U81504.1 | - | 4 - 5 | 468 | 106 | |
U91931.1 | - | 4 - 5 | 429 | 106 |