Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MMP27
Entrez Gene ID
64066
Gene Name
matrix metallopeptidase 27
Gene Aliases
MMP-27
Chromosome Location
Chr.11: 102690943 - 102705785 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022122.2 NP_071405.2 3 - 4 514 72
XM_011542949.2 XP_011541251.1 3 - 4 562 72
XM_011542950.2 XP_011541252.1 2 - 3 260 72
XM_017018120.1 XP_016873609.1 3 - 4 562 72
GenBank mRNA AF195192.1 - 3 - 4 497 72
AY358752.1 - 3 - 4 514 72

View Full Product Details