Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MMP27
Entrez Gene ID
64066
Gene Name
matrix metallopeptidase 27
Gene Aliases
MMP-27
Chromosome Location
Chr.11: 102690943 - 102705785 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022122.2 NP_071405.2 4 - 5 641 83
XM_011542950.2 XP_011541252.1 3 - 4 387 83
XM_017018120.1 XP_016873609.1 4 - 5 689 83
GenBank mRNA AF195192.1 - 4 - 5 624 83
AY358752.1 - 4 - 5 641 83

View Full Product Details