Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ELSPBP1
Entrez Gene ID
64100
Gene Name
epididymal sperm binding protein 1
Gene Aliases
E12, EDDM12, EL149, HE12
Chromosome Location
Chr.19: 47994651 - 48025153 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022142.4 NP_071425.3 6 - 7 855 96
XM_017027130.1 XP_016882619.1 6 - 7 954 96
GenBank mRNA AF279147.1 - 6 - 7 855 96
AJ278478.1 - 5 - 7 102
BC015598.2 - 6 - 7 819 96
BM661759.1 - 1 - 2 96
BM663735.1 - 1 - 2 96
GQ149084.1 - 6 - 7 819 96

View Full Product Details