Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ADAMTSL1
Entrez Gene ID
92949
Gene Name
ADAMTS like 1
Gene Aliases
ADAMTSL-1, ADAMTSR1, C9orf94, PUNCTIN
Chromosome Location
Chr.9: 17906574 - 18910950 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001040272.5 NP_001035362.3 22 - 23 4264 83
XM_011518063.2 XP_011516365.1 24 - 25 4402 83
XM_011518064.2 XP_011516366.1 23 - 24 4517 83
XM_011518067.1 XP_011516369.1 14 - 15 3132 83
XM_011518068.2 XP_011516370.1 12 - 13 3735 83
XM_011518070.2 XP_011516372.1 12 - 13 2900 83
XM_017015310.1 XP_016870799.1 23 - 24 11039 83
XM_017015311.1 XP_016870800.1 23 - 24 4351 83
XM_017015312.1 XP_016870801.1 22 - 23 4467 83
XM_017015313.1 XP_016870802.1 23 - 24 4165 83
XM_017015314.1 XP_016870803.1 24 - 25 4402 83
GenBank mRNA AK092602.1 - 1 - 2 323 83
BC100788.2 - 1 - 2 313 83
BC100789.2 - 1 - 2 315 83
BC100790.2 - 1 - 2 312 83
BX640951.1 - 3 - 4 397 83

View Full Product Details