The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001324458.1 | NP_001311387.1 | 3 - 4 | 220 | 60 |
NM_001324459.1 | NP_001311388.1 | 3 - 4 | 248 | 60 | |
NM_013292.4 | NP_037424.2 | 1 - 2 | 83 | 60 | |
GenBank mRNA | AF363061.1 | - | 1 - 2 | 56 | 60 |
AK314082.1 | - | 1 - 2 | 83 | 60 | |
BC012571.1 | - | 1 - 2 | 61 | 60 | |
BI828249.1 | - | 3 - 4 | 249 | 60 | |
BX649054.1 | - | 1 - 2 | 59 | 60 |