Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- CCDC91
- Entrez Gene ID
-
55297
- Gene Name
- coiled-coil domain containing 91
- Gene Aliases
-
HSD8, p56
- Chromosome Location
-
Chr.12: 28190427 - 28550166 on Build GRCh38
View Full Product Details