Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SFI1
Entrez Gene ID
9814
Gene Name
SFI1 centrin binding protein
Gene Aliases
PISD, PPP1R139, hSfi1p
Chromosome Location
Chr.22: 31496139 - 31618554 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007467.2 NP_001007468.1 17 - 18 2337 66
NM_001258325.1 NP_001245254.1 15 - 16 2172 66
NM_001258326.1 NP_001245255.1 15 - 16 2087 66
NM_001258327.1 NP_001245256.1 15 - 16 2091 66
NM_014775.3 NP_055590.2 16 - 17 2244 66
XM_005261868.2 XP_005261925.1 17 - 18 2335 66
XM_006724390.2 XP_006724453.1 15 - 16 2089 66
XM_011530570.2 XP_011528872.1 17 - 18 2335 66
XM_011530571.2 XP_011528873.1 17 - 18 2331 66
XM_011530574.2 XP_011528876.1 17 - 18 2012 66
XM_011530575.2 XP_011528877.1 17 - 18 2335 66
XM_011530576.2 XP_011528878.1 16 - 17 2263 66
XM_011530577.2 XP_011528879.1 16 - 17 2259 66
XM_011530579.2 XP_011528881.1 17 - 18 2012 66
XM_011530580.2 XP_011528882.1 16 - 17 2242 66
XM_011530581.2 XP_011528883.1 17 - 18 2012 66
XM_011530582.2 XP_011528884.1 15 - 16 2089 66
XM_011530583.2 XP_011528885.1 15 - 16 2085 66
XM_017029139.1 XP_016884628.1 16 - 17 1919 66
XM_017029140.1 XP_016884629.1 15 - 16 2170 66
XM_017029141.1 XP_016884630.1 16 - 17 1919 66
XM_017029142.1 XP_016884631.1 16 - 17 2263 66
XM_017029143.1 XP_016884632.1 16 - 17 2238 66
XM_017029144.1 XP_016884633.1 16 - 17 2242 66
XM_017029145.1 XP_016884634.1 15 - 16 2170 66
XM_017029147.1 XP_016884636.1 15 - 16 2170 66
XM_017029148.1 XP_016884637.1 15 - 16 2166 66
XM_017029149.1 XP_016884638.1 16 - 17 2263 66
XM_017029150.1 XP_016884639.1 16 - 17 2259 66
XM_017029151.1 XP_016884640.1 16 - 17 2238 66
XM_017029152.1 XP_016884641.1 14 - 15 1996 66
XM_017029153.1 XP_016884642.1 15 - 16 2085 66
XM_017029154.1 XP_016884643.1 15 - 16 2166 66
XM_017029155.1 XP_016884644.1 14 - 15 1996 66
XM_017029156.1 XP_016884645.1 14 - 15 1996 66
XM_017029157.1 XP_016884646.1 13 - 14 1498 66
XM_017029158.1 XP_016884647.1 17 - 18 2331 66
GenBank mRNA AB011114.2 - 16 - 17 2108 66
AK292408.1 - 17 - 18 1913 66
AK293987.1 - 15 - 16 1785 66
AK303362.1 - 15 - 16 1722 66
BC110814.1 - 15 - 16 1688 66
BC129926.1 - 13 - 14 1630 66
BC129945.1 - 16 - 17 2017 66
DA391957.1 - 3 - 4 382 66
DA534059.1 - 3 - 4 327 66

View Full Product Details