Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SFI1
Entrez Gene ID
9814
Gene Name
SFI1 centrin binding protein
Gene Aliases
PISD, PPP1R139, hSfi1p
Chromosome Location
Chr.22: 31496139 - 31618554 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007467.2 NP_001007468.1 27 - 28 3525 121
NM_001258325.1 NP_001245254.1 25 - 26 3360 121
NM_001258326.1 NP_001245255.1 25 - 26 3275 121
NM_001258327.1 NP_001245256.1 25 - 26 3243 121
NM_014775.3 NP_055590.2 26 - 27 3432 121
XM_005261868.2 XP_005261925.1 27 - 28 3523 121
XM_006724390.2 XP_006724453.1 25 - 26 3277 121
XM_011530570.2 XP_011528872.1 28 - 29 3574 121
XM_011530571.2 XP_011528873.1 28 - 29 3570 121
XM_011530574.2 XP_011528876.1 28 - 29 3251 121
XM_011530575.2 XP_011528877.1 27 - 28 3523 121
XM_011530576.2 XP_011528878.1 27 - 28 3502 121
XM_011530577.2 XP_011528879.1 27 - 28 3498 121
XM_011530579.2 XP_011528881.1 28 - 29 3251 121
XM_011530580.2 XP_011528882.1 27 - 28 3481 121
XM_011530581.2 XP_011528883.1 28 - 29 3251 121
XM_011530582.2 XP_011528884.1 26 - 27 3328 121
XM_011530583.2 XP_011528885.1 26 - 27 3324 121
XM_011530587.2 XP_011528889.1 17 - 18 1948 121
XM_017029139.1 XP_016884628.1 26 - 27 3107 121
XM_017029140.1 XP_016884629.1 26 - 27 3409 121
XM_017029141.1 XP_016884630.1 27 - 28 3158 121
XM_017029142.1 XP_016884631.1 26 - 27 3451 121
XM_017029143.1 XP_016884632.1 26 - 27 3426 121
XM_017029144.1 XP_016884633.1 26 - 27 3430 121
XM_017029145.1 XP_016884634.1 25 - 26 3358 121
XM_017029146.1 XP_016884635.1 26 - 27 2990 121
XM_017029147.1 XP_016884636.1 25 - 26 3358 121
XM_017029148.1 XP_016884637.1 25 - 26 3354 121
XM_017029149.1 XP_016884638.1 26 - 27 3451 121
XM_017029150.1 XP_016884639.1 26 - 27 3447 121
XM_017029151.1 XP_016884640.1 26 - 27 3426 121
XM_017029152.1 XP_016884641.1 25 - 26 3235 121
XM_017029153.1 XP_016884642.1 25 - 26 3273 121
XM_017029154.1 XP_016884643.1 25 - 26 3354 121
XM_017029155.1 XP_016884644.1 24 - 25 3184 121
XM_017029156.1 XP_016884645.1 24 - 25 3184 121
XM_017029157.1 XP_016884646.1 24 - 25 2737 121
XM_017029159.1 XP_016884648.1 23 - 24 2749 121
XM_017029160.1 XP_016884649.1 15 - 16 1684 121
XM_017029161.1 XP_016884650.1 23 - 24 2785 121
XM_017029162.1 XP_016884651.1 23 - 24 2785 121
GenBank mRNA AB011114.2 - 26 - 27 3296 121
AK055147.1 - 15 - 16 1764 121
AK292408.1 - 27 - 28 3101 121
AK293987.1 - 25 - 26 2973 121
AK303362.1 - 25 - 26 2874 121
BC021576.1 - 9 - 10 1092 121
BC110814.1 - 25 - 26 2876 121
BC129926.1 - 23 - 24 2818 121
BC129945.1 - 21 - 22 2524 121

View Full Product Details