The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152526.5 | NP_689739.4 | 10 - 11 | 1641 | 70 |
XM_017003285.1 | XP_016858774.1 | 12 - 13 | 6714 | 70 | |
GenBank mRNA | AF428251.1 | - | 10 - 11 | 1434 | 70 |