Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AOX1
Entrez Gene ID
316
Gene Name
aldehyde oxidase 1
Gene Aliases
AO, AOH1
Chromosome Location
Chr.2: 200585952 - 200677064 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001159.3 NP_001150.3 30 - 31 3527 87
XM_011511062.1 XP_011509364.1 30 - 31 3583 87
XM_017003946.1 XP_016859435.1 30 - 31 3583 87
XM_017003947.1 XP_016859436.1 30 - 31 3583 87
GenBank mRNA AB046692.1 - 30 - 31 3526 87
AK297930.1 - 18 - 19 2358 87
AK307738.1 - 17 - 18 87
BC117179.1 - 30 - 31 3480 87
BC117181.1 - 30 - 31 3480 87
L11005.1 - 30 - 31 3556 87

View Full Product Details