Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC39A10
Entrez Gene ID
57181
Gene Name
solute carrier family 39 member 10
Gene Aliases
LZT-Hs2
Chromosome Location
Chr.2: 195575949 - 195737702 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001127257.1 NP_001120729.1 1 - 2 196 111
GenBank mRNA BC029413.1 - 1 - 2 257 111
CR749813.1 - 1 - 2 47 111

View Full Product Details