Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NDUFS1
Entrez Gene ID
4719
Gene Name
NADH:ubiquinone oxidoreductase core subunit S1
Gene Aliases
CI-75Kd, CI-75k, PRO1304
Chromosome Location
Chr.2: 206121971 - 206159519 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199981.1 NP_001186910.1 15 - 16 1963 101
NM_001199982.1 NP_001186911.1 13 - 14 1794 101
NM_001199983.1 NP_001186912.1 15 - 16 1979 101
NM_001199984.1 NP_001186913.1 16 - 17 1991 101
NM_005006.6 NP_004997.4 16 - 17 2071 101
XM_017004188.1 XP_016859677.1 16 - 17 2030 101
GenBank mRNA AK098395.1 - 11 - 12 1295 101
AK295705.1 - 15 - 16 1869 101
AK295966.1 - 12 - 13 1635 101
AK295987.1 - 16 - 17 1991 101
AK298320.1 - 13 - 14 1700 101
AK300585.1 - 15 - 16 1883 101
BC012068.1 - 4 - 5 577 101
BC018900.1 - 16 - 17 1947 101
BC022368.1 - 16 - 17 1972 101
BC030833.1 - 16 - 17 1975 101
X61100.1 - 16 - 17 1934 101

View Full Product Details