The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001199981.1 | NP_001186910.1 | 16 - 17 | 2090 | 70 |
NM_001199982.1 | NP_001186911.1 | 14 - 15 | 1921 | 70 | |
NM_001199983.1 | NP_001186912.1 | 16 - 17 | 2106 | 70 | |
NM_001199984.1 | NP_001186913.1 | 17 - 18 | 2118 | 70 | |
NM_005006.6 | NP_004997.4 | 17 - 18 | 2198 | 70 | |
XM_017004188.1 | XP_016859677.1 | 17 - 18 | 2157 | 70 | |
GenBank mRNA | AK098395.1 | - | 12 - 13 | 1422 | 70 |
AK295705.1 | - | 16 - 17 | 1996 | 70 | |
AK295966.1 | - | 13 - 14 | 1762 | 70 | |
AK295987.1 | - | 17 - 18 | 2118 | 70 | |
AK298320.1 | - | 14 - 15 | 1827 | 70 | |
AK300585.1 | - | 16 - 17 | 2010 | 70 | |
BC012068.1 | - | 5 - 6 | 704 | 70 | |
BC018900.1 | - | 17 - 18 | 2074 | 70 | |
BC022368.1 | - | 17 - 18 | 2099 | 70 | |
BC030833.1 | - | 17 - 18 | 2102 | 70 | |
X61100.1 | - | 17 - 18 | 2061 | 70 |