The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018410.4 | NP_060880.3 | 5 - 6 | 465 | 117 |
GenBank mRNA | AB101211.1 | - | 5 - 6 | 466 | 117 |
AB162218.1 | - | 5 - 6 | 399 | 117 | |
AK074809.1 | - | 5 - 6 | 431 | 117 | |
BC001940.1 | - | 5 - 6 | 399 | 117 | |
BE535238.1 | - | 3 - 4 | 230 | 117 | |
BM838206.1 | - | 6 - 7 | 432 | 117 | |
BQ225710.1 | - | 5 - 6 | 421 | 118 | |
DB022872.1 | - | 5 - 6 | 463 | 117 |