The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014640.4 | NP_055455.3 | 18 - 19 | 3629 | 103 |
XM_006712875.2 | XP_006712938.1 | 18 - 19 | 3639 | 103 | |
XM_017005392.1 | XP_016860881.1 | 16 - 17 | 3794 | 103 | |
GenBank mRNA | AK291770.1 | - | 18 - 19 | 3587 | 103 |
AK302728.1 | - | 18 - 19 | 3175 | 103 | |
AL833511.1 | - | 18 - 19 | 3278 | 103 | |
BC021707.2 | - | 18 - 19 | 3554 | 103 | |
D79995.1 | - | 18 - 19 | 3466 | 103 |