The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005199.4 | NP_005190.4 | 1 - 2 | 79 | 122 |
GenBank mRNA | AK125362.1 | - | 1 - 2 | 79 | 122 |
AK309340.1 | - | 1 - 2 | 79 | 122 | |
BC111802.1 | - | 1 - 2 | 58 | 122 |