The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001166269.1 | NP_001159741.1 | 8 - 9 | 1090 | 75 |
NM_001166270.1 | NP_001159742.1 | 7 - 8 | 947 | 75 | |
NM_017815.2 | NP_060285.2 | 8 - 9 | 1082 | 75 | |
GenBank mRNA | AK000431.1 | - | 8 - 9 | 1051 | 75 |
AK026016.1 | - | 8 - 9 | 1036 | 75 | |
AK057779.1 | - | 6 - 7 | 877 | 75 | |
AY927599.1 | - | 3 - 4 | 330 | 75 | |
BC001916.1 | - | 8 - 9 | 1079 | 75 | |
BC002554.2 | - | 8 - 9 | 1013 | 75 | |
BX247981.1 | - | 5 - 6 | 888 | 75 | |
BX247995.1 | - | 7 - 8 | 899 | 75 | |
BX248256.1 | - | 7 - 8 | 914 | 75 |