The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000492.3 | NP_000483.3 | 3 - 4 | 406 | 140 |
XM_011515751.2 | XP_011514053.1 | 4 - 5 | 379 | 140 | |
XM_011515753.1 | XP_011514055.1 | 4 - 5 | 496 | 140 | |
XM_011515754.2 | XP_011514056.1 | 6 - 7 | 1471 | 140 | |
XM_017011699.1 | XP_016867188.1 | 5 - 6 | 1217 | 140 | |
GenBank mRNA | M28668.1 | - | 3 - 4 | 406 | 140 |