The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019071.2 | NP_061944.2 | 8 - 9 | 850 | 139 |
GenBank mRNA | AF074968.1 | - | 8 - 9 | 827 | 139 |
AF180298.1 | - | 8 - 9 | 817 | 139 | |
AK000096.1 | - | 8 - 9 | 851 | 139 | |
AK293682.1 | - | 7 - 8 | 736 | 139 | |
AL603623.1 | - | 8 - 9 | 827 | 139 | |
AY007790.1 | - | 8 - 9 | 831 | 139 | |
BC010851.1 | - | 7 - 8 | 700 | 139 |