The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000245.3 | NP_000236.2 | 6 - 7 | 2063 | 73 |
NM_001127500.2 | NP_001120972.1 | 6 - 7 | 2063 | 73 | |
NM_001324401.1 | NP_001311330.1 | 6 - 7 | 2063 | 73 | |
NM_001324402.1 | NP_001311331.1 | 5 - 6 | 880 | 73 | |
XM_006715990.2 | XP_006716053.1 | 6 - 7 | 802 | 73 | |
XM_011516223.1 | XP_011514525.1 | 7 - 8 | 2092 | 73 | |
GenBank mRNA | AB209898.1 | - | 5 - 6 | 838 | 73 |
AK296974.1 | - | 6 - 7 | 780 | 73 | |
BC130420.1 | - | 6 - 7 | 2026 | 73 | |
EU176015.1 | - | 6 - 7 | 2055 | 73 | |
EU826567.1 | - | 5 - 6 | 1861 | 73 | |
EU826568.1 | - | 5 - 6 | 1861 | 73 | |
EU826570.1 | - | 5 - 6 | 1861 | 73 | |
EU826571.1 | - | 5 - 6 | 1861 | 73 | |
EU826572.1 | - | 5 - 6 | 1861 | 73 | |
EU826573.1 | - | 5 - 6 | 1861 | 73 | |
EU826574.1 | - | 5 - 6 | 1777 | 73 | |
EU826575.1 | - | 5 - 6 | 1861 | 73 | |
J02958.1 | - | 6 - 7 | 2055 | 73 | |
X54559.1 | - | 6 - 7 | 2055 | 73 |