Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SPRYD3
Entrez Gene ID
84926
Gene Name
SPRY domain containing 3
Gene Aliases
-
Chromosome Location
Chr.12: 53064316 - 53079420 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_032840.2 NP_116229.1 3 - 4 330 76
GenBank mRNA AK027706.1 - 3 - 4 264 76
AK074694.1 - 3 - 4 296 76
AK294092.1 - 3 - 4 314 76
BC136312.1 - 3 - 4 263 76
DA389433.1 - 3 - 4 330 76

View Full Product Details