Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SPRYD3
Entrez Gene ID
84926
Gene Name
SPRY domain containing 3
Gene Aliases
-
Chromosome Location
Chr.12: 53064316 - 53079420 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_032840.2 NP_116229.1 5 - 6 596 61
GenBank mRNA AK027706.1 - 5 - 6 530 61
AK074694.1 - 5 - 6 562 61
BC136312.1 - 5 - 6 529 61

View Full Product Details