The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032840.2 | NP_116229.1 | 8 - 9 | 983 | 94 |
GenBank mRNA | AK027706.1 | - | 8 - 9 | 917 | 94 |
AK074566.1 | - | 3 - 4 | 267 | 94 | |
AK074694.1 | - | 8 - 9 | 949 | 94 | |
AK294092.1 | - | 7 - 8 | 831 | 94 | |
BC136312.1 | - | 8 - 9 | 916 | 94 |