The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001307924.1 | NP_001294853.1 | 1 - 2 | 468 | 95 |
NM_004537.5 | NP_004528.1 | 1 - 2 | 468 | 95 | |
NM_139207.3 | NP_631946.1 | 1 - 2 | 468 | 95 | |
XM_006719425.3 | XP_006719488.1 | 1 - 2 | 120 | 95 | |
XM_011538390.2 | XP_011536692.2 | 1 - 2 | 127 | 95 | |
XM_017019340.1 | XP_016874829.1 | 1 - 2 | 120 | 95 | |
GenBank mRNA | AK304787.1 | - | 1 - 2 | 170 | 95 |
AL162068.1 | - | 1 - 2 | 105 | 95 | |
BC002387.2 | - | 1 - 2 | 102 | 95 | |
BU183927.1 | - | - | 467 | 95 | |
DC300275.1 | - | 1 - 2 | 412 | 95 | |
DC321631.1 | - | 1 - 2 | 394 | 95 |