Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
VEZT
Entrez Gene ID
55591
Gene Name
vezatin, adherens junctions transmembrane protein
Gene Aliases
VEZATIN
Chromosome Location
Chr.12: 95217746 - 95302799 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK001338.1 - 6 - 7 827 109

View Full Product Details