The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001251904.1 | NP_001238833.1 | 17 - 18 | 1868 | 79 |
NM_001251905.1 | NP_001238834.1 | 17 - 18 | 1893 | 79 | |
NM_018171.3 | NP_060641.2 | 17 - 18 | 1850 | 79 | |
XM_006719472.1 | XP_006719535.1 | 17 - 18 | 1787 | 79 | |
XM_011538530.2 | XP_011536832.1 | 17 - 18 | 3320 | 79 | |
XM_011538531.2 | XP_011536833.1 | 17 - 18 | 2983 | 79 | |
XM_011538532.2 | XP_011536834.1 | 17 - 18 | 4309 | 79 | |
XM_017019551.1 | XP_016875040.1 | 17 - 18 | 2255 | 79 | |
XM_017019552.1 | XP_016875041.1 | 17 - 18 | 2965 | 79 | |
XM_017019553.1 | XP_016875042.1 | 18 - 19 | 4563 | 79 | |
XM_017019554.1 | XP_016875043.1 | 17 - 18 | 1769 | 79 | |
GenBank mRNA | AK001521.2 | - | 17 - 18 | 79 | |
AK290562.1 | - | 17 - 18 | 1703 | 79 | |
AK293783.1 | - | 17 - 18 | 1642 | 79 | |
AK296610.1 | - | 17 - 18 | 1649 | 79 | |
AK297100.1 | - | 17 - 18 | 1721 | 79 | |
AK310377.1 | - | 17 - 18 | 1638 | 79 | |
AY113704.1 | - | 17 - 18 | 1850 | 79 | |
BC033731.1 | - | 17 - 18 | 1754 | 79 |