The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001251904.1 | NP_001238833.1 | 18 - 19 | 1906 | 94 |
NM_001251905.1 | NP_001238834.1 | 18 - 19 | 1931 | 94 | |
NM_018171.3 | NP_060641.2 | 18 - 19 | 1888 | 94 | |
XM_011538530.2 | XP_011536832.1 | 18 - 19 | 3358 | 94 | |
XM_011538531.2 | XP_011536833.1 | 18 - 19 | 3021 | 94 | |
XM_011538532.2 | XP_011536834.1 | 18 - 19 | 4347 | 94 | |
XM_017019551.1 | XP_016875040.1 | 18 - 19 | 2293 | 94 | |
XM_017019552.1 | XP_016875041.1 | 18 - 19 | 3003 | 94 | |
XM_017019553.1 | XP_016875042.1 | 19 - 20 | 4601 | 94 | |
GenBank mRNA | AK290562.1 | - | 18 - 19 | 1741 | 94 |
AK296610.1 | - | 18 - 19 | 1687 | 94 | |
AK297100.1 | - | 18 - 19 | 1759 | 94 | |
AY113704.1 | - | 18 - 19 | 1888 | 94 | |
BC033731.1 | - | 18 - 19 | 1792 | 94 |