Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GNPTAB
Entrez Gene ID
79158
Gene Name
N-acetylglucosamine-1-phosphate transferase alpha and beta subunits
Gene Aliases
GNPTA, ICD
Chromosome Location
Chr.12: 101745497 - 101830867 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024312.4 NP_077288.2 4 - 5 561 93
XM_006719593.2 XP_006719656.1 4 - 5 556 93
XM_011538731.2 XP_011537033.1 4 - 5 557 93
XM_017019961.1 XP_016875450.1 4 - 5 458 93
XM_017019962.1 XP_016875451.1 4 - 5 540 93
GenBank mRNA AK056137.1 - 4 - 5 432 93
AM085438.1 - 4 - 5 548 93
AY687932.1 - 4 - 5 533 93
BC071687.1 - 4 - 5 509 93
BC131787.1 - 4 - 5 489 93

View Full Product Details