The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001178003.1 | NP_001171474.1 | 2 - 3 | 547 | 91 |
NM_144668.5 | NP_653269.3 | 2 - 3 | 534 | 91 | |
GenBank mRNA | AK097102.1 | - | 1 - 2 | 288 | 91 |
BC028421.2 | - | 2 - 3 | 532 | 91 | |
BC036233.1 | - | 2 - 3 | 527 | 91 | |
BG717296.1 | - | 2 - 3 | 567 | 91 |