The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_104219.1 | 1 - 2 | 748 | 81 | |
XM_005268576.4 | XP_005268633.1 | 1 - 2 | 2001 | 81 | |
XM_006719199.2 | XP_006719262.1 | 1 - 2 | 760 | 81 | |
XM_011537774.2 | XP_011536076.1 | 1 - 2 | 760 | 81 | |
XM_017018716.1 | XP_016874205.1 | 1 - 2 | 760 | 81 | |
GenBank mRNA | AK098214.1 | - | 1 - 2 | 748 | 81 |