Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
XRRA1
Entrez Gene ID
143570
Gene Name
X-ray radiation resistance associated 1
Gene Aliases
-
Chromosome Location
Chr.11: 74840910 - 74949187 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001270381.1 NP_001257310.1 15 - 16 2193 68
NM_182969.2 NP_892014.1 18 - 19 2461 68
XM_005273765.2 XP_005273822.1 18 - 19 2485 68
XM_005273770.2 XP_005273827.1 16 - 17 2265 68
XM_011544755.1 XP_011543057.1 18 - 19 2440 68
XM_011544756.1 XP_011543058.1 17 - 18 2359 68
XM_011544757.1 XP_011543059.1 17 - 18 2314 68
XM_011544758.1 XP_011543060.1 16 - 17 2314 68
XM_011544759.1 XP_011543061.1 16 - 17 2290 68
XM_011544761.1 XP_011543063.1 15 - 16 2242 68
XM_011544762.1 XP_011543064.1 15 - 16 2224 68
XM_011544764.1 XP_011543066.1 15 - 16 2149 68
XM_011544765.1 XP_011543067.1 16 - 17 2199 68
XM_011544766.2 XP_011543068.1 18 - 19 2504 68
XM_011544767.1 XP_011543069.1 17 - 18 2406 68
XM_011544768.1 XP_011543070.1 16 - 17 2253 68
XM_017017221.1 XP_016872710.1 16 - 17 2275 68
XM_017017226.1 XP_016872715.1 17 - 18 2380 68
XM_017017227.1 XP_016872716.1 17 - 18 2356 68
XM_017017229.1 XP_016872718.1 16 - 17 2209 68
XM_017017230.1 XP_016872719.1 15 - 16 2033 68
XM_017017231.1 XP_016872720.1 14 - 15 2119 68
XM_017017233.1 XP_016872722.1 15 - 16 2213 68
XM_017017234.1 XP_016872723.1 14 - 15 2098 68
GenBank mRNA AF439934.1 - 7 - 8 1086 68
AK074152.1 - 13 - 14 1791 68
BC037294.2 - 15 - 16 2126 68
BC040541.1 - 15 - 16 2126 68

View Full Product Details