The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001300869.1 | NP_001287798.1 | 2 - 3 | 372 | 107 |
XM_011544974.1 | XP_011543276.1 | 2 - 3 | 357 | 107 | |
XM_017017632.1 | XP_016873121.1 | 2 - 3 | 357 | 107 | |
GenBank mRNA | BC011387.1 | - | 1 - 2 | 87 | 107 |
DB124589.1 | - | 2 - 3 | 358 | 107 |