Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZEB1
Entrez Gene ID
6935
Gene Name
zinc finger E-box binding homeobox 1
Gene Aliases
AREB6, BZP, DELTAEF1, FECD6, NIL2A, PPCD3, TCF8, ZFHEP, ZFHX1A
Chromosome Location
Chr.10: 31318495 - 31529814 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001128128.2 NP_001121600.1 4 - 5 829 64
NM_001174093.1 NP_001167564.1 3 - 4 489 64
NM_001174094.1 NP_001167565.1 4 - 5 826 64
NM_001174095.1 NP_001167566.1 3 - 4 348 64
NM_001174096.1 NP_001167567.1 4 - 5 552 64
NM_001323638.1 NP_001310567.1 8 - 9 1199 64
NM_001323641.1 NP_001310570.1 4 - 5 779 64
NM_001323642.1 NP_001310571.1 6 - 7 1081 64
NM_001323643.1 NP_001310572.1 6 - 7 859 64
NM_001323644.1 NP_001310573.1 3 - 4 578 64
NM_001323645.1 NP_001310574.1 5 - 6 922 64
NM_001323646.1 NP_001310575.1 7 - 8 1160 64
NM_001323647.1 NP_001310576.1 5 - 6 692 64
NM_001323648.1 NP_001310577.1 4 - 5 632 64
NM_001323649.1 NP_001310578.1 5 - 6 695 64
NM_001323650.1 NP_001310579.1 7 - 8 927 64
NM_001323651.1 NP_001310580.1 5 - 6 595 64
NM_001323652.1 NP_001310581.1 6 - 7 744 64
NM_001323653.1 NP_001310582.1 6 - 7 851 64
NM_001323654.1 NP_001310583.1 6 - 7 738 64
NM_001323655.1 NP_001310584.1 6 - 7 848 64
NM_001323656.1 NP_001310585.1 4 - 5 523 64
NM_001323657.1 NP_001310586.1 4 - 5 526 64
NM_001323658.1 NP_001310587.1 4 - 5 365 64
NM_001323659.1 NP_001310588.1 4 - 5 583 64
NM_001323660.1 NP_001310589.1 4 - 5 580 64
NM_001323661.1 NP_001310590.1 5 - 6 726 64
NM_001323662.1 NP_001310591.1 6 - 7 1128 64
NM_001323663.1 NP_001310592.1 7 - 8 1067 64
NM_001323664.1 NP_001310593.1 6 - 7 1051 64
NM_001323665.1 NP_001310594.1 5 - 6 969 64
NM_001323666.1 NP_001310595.1 4 - 5 909 64
NM_001323671.1 NP_001310600.1 5 - 6 805 64
NM_001323672.1 NP_001310601.1 6 - 7 857 64
NM_001323673.1 NP_001310602.1 5 - 6 802 64
NM_001323676.1 NP_001310605.1 4 - 5 662 64
NM_001323677.1 NP_001310606.1 4 - 5 659 64
NM_030751.5 NP_110378.3 4 - 5 549 64
XM_006717498.2 XP_006717561.1 7 - 8 987 64
XM_011519643.2 XP_011517945.1 7 - 8 1290 64
XM_017016597.1 XP_016872086.1 7 - 8 889 64
XM_017016598.1 XP_016872087.1 7 - 8 4019 64
XM_017016599.1 XP_016872088.1 6 - 7 3857 64
XM_017016600.1 XP_016872089.1 8 - 9 4098 64
XM_017016601.1 XP_016872090.1 7 - 8 4016 64
XM_017016602.1 XP_016872091.1 8 - 9 4095 64
XM_017016603.1 XP_016872092.1 7 - 8 3933 64
GenBank mRNA AK091478.1 - 3 - 4 449 64
AK294775.1 - 4 - 5 744 64
AK296244.1 - 4 - 5 472 64
AK300830.1 - 3 - 4 298 64
AK307510.1 - 5 - 6 555 64
AK308785.1 - 5 - 6 634 64
AK308822.1 - 6 - 7 694 64
AK309399.1 - 5 - 6 667 64
AK310924.1 - 5 - 6 656 64
AK311116.1 - 4 - 5 562 64
AK311187.1 - 4 - 5 812 64
AK311201.1 - 4 - 5 776 64
AK311372.1 - 7 - 8 1106 64
AK311557.1 - 6 - 7 1132 64
AK314683.1 - 4 - 5 510 64
AL831979.1 - 6 - 7 807 64
BC009572.1 - 4 - 5 505 64
BC065705.1 - 4 - 5 510 64
BC107781.1 - 4 - 5 505 64
BC112392.1 - 4 - 5 505 64
BX647794.1 - 8 - 9 1200 64
DC299346.1 - 4 - 5 525 64
DC403144.1 - 4 - 5 365 64
U12170.1 - 4 - 5 510 64

View Full Product Details