The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172236.1 | NP_758436.1 | 4 - 5 | 592 | 85 |
GenBank mRNA | BC000582.2 | - | 4 - 5 | 592 | 85 |