Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BPIFA1
Entrez Gene ID
51297
Gene Name
BPI fold containing family A member 1
Gene Aliases
LUNX, NASG, PLUNC, SPLUNC1, SPURT, bA49G10.5
Chromosome Location
Chr.20: 33235996 - 33243309 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001243193.1 NP_001230122.1 6 - 7 733 90
NM_016583.3 NP_057667.1 6 - 7 733 90
NM_130852.2 NP_570913.1 6 - 7 733 90
GenBank mRNA AB024937.1 - 6 - 7 730 90
AF158745.1 - 6 - 7 724 90
AF417256.1 - 6 - 7 733 90
AF417257.1 - 6 - 7 733 90
AF439448.1 - 6 - 7 717 90
AK292778.1 - 6 - 7 732 90
AY359020.1 - 6 - 7 734 90
BC012549.1 - 6 - 7 737 90
DQ846869.1 - 4 - 5 621 90

View Full Product Details