The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001184699.1 | NP_001171628.1 | 2 - 3 | 410 | 57 |
NM_019609.4 | NP_062555.1 | 2 - 3 | 410 | 57 | |
GenBank mRNA | AK027661.1 | - | 2 - 3 | 346 | 57 |
AK075527.1 | - | 2 - 3 | 385 | 57 | |
AK315027.1 | - | 2 - 3 | 397 | 57 | |
AL541188.3 | - | 2 - 3 | 382 | 57 | |
AY358956.1 | - | 2 - 3 | 366 | 57 | |
BC063430.1 | - | 2 - 3 | 387 | 57 | |
BX366559.2 | - | 2 - 3 | 392 | 57 | |
CN308423.1 | - | 2 - 3 | 401 | 57 |