The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001135934.1 | NP_001129406.1 | 7 - 8 | 1009 | 110 |
NM_001135935.1 | NP_001129407.1 | 7 - 8 | 1009 | 110 | |
NM_001135936.1 | NP_001129408.1 | 7 - 8 | 1009 | 110 | |
NM_001286665.1 | NP_001273594.1 | 7 - 8 | 1009 | 110 | |
NM_001286666.1 | NP_001273595.1 | 7 - 8 | 1009 | 110 | |
NM_001286667.1 | NP_001273596.1 | 7 - 8 | 1009 | 110 | |
NM_006475.2 | NP_006466.2 | 7 - 8 | 1009 | 110 | |
XM_005266231.2 | XP_005266288.1 | 7 - 8 | 1004 | 110 | |
XM_005266232.2 | XP_005266289.1 | 7 - 8 | 1004 | 110 | |
XM_017020355.1 | XP_016875844.1 | 7 - 8 | 1004 | 110 | |
XM_017020356.1 | XP_016875845.1 | 7 - 8 | 1004 | 110 | |
GenBank mRNA | AK300261.1 | - | 7 - 8 | 1123 | 110 |
AY140646.1 | - | 7 - 8 | 902 | 110 | |
AY918092.1 | - | 7 - 8 | 902 | 110 | |
BC106709.1 | - | 7 - 8 | 902 | 110 | |
BC106710.1 | - | 7 - 8 | 902 | 110 | |
D13665.1 | - | 7 - 8 | 918 | 110 | |
D13666.1 | - | 7 - 8 | 902 | 110 | |
EU262883.1 | - | 7 - 8 | 891 | 110 | |
EU262884.1 | - | 7 - 8 | 891 | 110 | |
EU262885.1 | - | 7 - 8 | 891 | 110 | |
EU262886.1 | - | 7 - 8 | 891 | 110 | |
GU354210.1 | - | 7 - 8 | 902 | 110 |