Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EPSTI1
Entrez Gene ID
94240
Gene Name
epithelial stromal interaction 1 (breast)
Gene Aliases
BRESI1
Chromosome Location
Chr.13: 42886388 - 42992271 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002264.2 NP_001002264.1 2 - 3 355 102
NM_033255.3 NP_150280.1 2 - 3 355 102
XM_005266596.1 XP_005266653.1 2 - 3 355 102
XM_005266597.1 XP_005266654.1 2 - 3 355 102
XM_006719896.3 XP_006719959.1 2 - 3 355 102
XM_011535312.1 XP_011533614.1 2 - 3 1565 102
XM_017020853.1 XP_016876342.1 2 - 3 145 102
XM_017020854.1 XP_016876343.1 3 - 4 731 102
GenBank mRNA AF396928.1 - 2 - 3 314 102
AK292461.1 - 2 - 3 325 102
BC023660.2 - 2 - 3 314 102
BP228419.1 - 3 - 4 324 102
BU603558.1 - - 328 102
DA940572.1 - 2 - 3 356 102

View Full Product Details