The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001002264.2 | NP_001002264.1 | 3 - 4 | 435 | 95 |
NM_033255.3 | NP_150280.1 | 3 - 4 | 435 | 95 | |
XM_005266596.1 | XP_005266653.1 | 3 - 4 | 435 | 95 | |
XM_005266597.1 | XP_005266654.1 | 3 - 4 | 435 | 95 | |
XM_006719896.3 | XP_006719959.1 | 3 - 4 | 435 | 95 | |
XM_011535312.1 | XP_011533614.1 | 3 - 4 | 1645 | 95 | |
XM_011535313.1 | XP_011533615.1 | 2 - 3 | 340 | 95 | |
XM_017020853.1 | XP_016876342.1 | 3 - 4 | 225 | 95 | |
XM_017020854.1 | XP_016876343.1 | 4 - 5 | 811 | 95 | |
XM_017020855.1 | XP_016876344.1 | 2 - 3 | 1024 | 95 | |
GenBank mRNA | AF396928.1 | - | 3 - 4 | 394 | 95 |
AK292461.1 | - | 3 - 4 | 405 | 95 | |
AK313877.1 | - | 3 - 4 | 399 | 95 | |
AL831953.1 | - | 2 - 3 | 1827 | 95 | |
BC023660.2 | - | 3 - 4 | 394 | 95 | |
BP228419.1 | - | 4 - 5 | 404 | 95 | |
BU603558.1 | - | - | 408 | 95 | |
DA940572.1 | - | 3 - 4 | 436 | 95 |