Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KIAA0226L
Entrez Gene ID
80183
Gene Name
KIAA0226 like
Gene Aliases
C13orf18
Chromosome Location
Chr.13: 46342000 - 46390042 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001286761.1 NP_001273690.1 5 - 6 1416 132
NM_001286762.1 NP_001273691.1 4 - 5 1322 132
NM_001286763.1 NP_001273692.1 5 - 6 1051 132
NM_001286764.1 NP_001273693.1 4 - 5 935 132
NM_001286765.1 NP_001273694.1 4 - 5 781 132
NM_001286766.1 NP_001273695.1 3 - 4 665 132
NM_025113.3 NP_079389.2 5 - 6 1438 132
GenBank mRNA AK025215.1 - 4 - 5 1071 132
AK093073.1 - 5 - 6 1386 132
AK125950.1 - 5 - 6 1423 132
AK293034.1 - 4 - 5 687 132
AK300183.1 - 4 - 5 485 132
AK307854.1 - 3 - 4 408 132
AK310264.1 - 6 - 7 1425 132
BC004495.1 - 1 - 2 135 132
BC032311.1 - 4 - 5 1003 132
BC043488.1 - 5 - 6 784 132
BX641126.1 - 3 - 4 472 132

View Full Product Details